Recombinat Protein,Mouse EpCAM TROP1 Protein, His Tag,Mouse EpCAM, His Tag (EPM-M5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Thr 266 (Accession AAH05618.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse EpCAM TROP1 Protein, His Tag,Mouse EpCAM, His Tag (EPM-M5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Thr 266 (Accession AAH05618.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse EpCAM TROP1 Protein, His Tag,Mouse EpCAM, His Tag (EPM-M5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Thr 266 (Accession AAH05618.1).This protein carries a polyhistidine tag at the C-terminus. The 1 * 1 mg ACRBEPM-M5227-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou