Recombinat Protein,Human GDF-15 MIC-1 Protein, His Tag,Human GDF-15, His Tag (GD5-H5149) is expressed from E.coli cells. It contain s AA Ala 197 - Ile 308 (Accession Q99988-1).This protein carries a polyhistidine tag at the N-terminus. The protein h 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human GDF-15 MIC-1 Protein, His Tag,Human GDF-15, His Tag (GD5-H5149) is expressed from E.coli cells. It contain s AA Ala 197 - Ile 308 (Accession Q99988-1).This protein carries a polyhistidine tag at the N-terminus. The protein h 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human GDF-15 MIC-1 Protein, His Tag,Human GDF-15, His Tag (GD5-H5149) is expressed from E.coli cells. It contain s AA Ala 197 - Ile 308 (Accession Q99988-1).This protein carries a polyhistidine tag at the N-terminus. The protein h 1 * 1 mg ACRBGD5-H5149-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou