MABSol® Biotinylated Human Her2, His Tag, primary amine labeling, long spacer (HE2-H822R) is expressed from human HEK293 cells. It contains AA Thr 23 - Thr 652 (Accession P04626-1). It is the biotinylated form of Human Her2, His Tag (Cat. No. HE2-H 1 * 200 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human Her2, His Tag, primary amine labeling, long spacer (HE2-H822R) is expressed from human HEK293 cells. It contains AA Thr 23 - Thr 652 (Accession P04626-1). It is the biotinylated form of Human Her2, His Tag (Cat. No. HE2-H 1 * 200 µG ACRBHE2-H822R200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou