Recombinat Protein,Mouse IL-13 R alpha 1 Protein, His Tag,Mouse IL-13 R alpha 1, His Tag (IL1-M52H7) is expressed from human 293 cel ls (HEK293). It contains AA Ala 26 - Thr 340 (Accession NP598751).This protein carries a polyhistidine tag at the C- 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse IL-13 R alpha 1 Protein, His Tag,Mouse IL-13 R alpha 1, His Tag (IL1-M52H7) is expressed from human 293 cel ls (HEK293). It contains AA Ala 26 - Thr 340 (Accession NP598751).This protein carries a polyhistidine tag at the C- 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse IL-13 R alpha 1 Protein, His Tag,Mouse IL-13 R alpha 1, His Tag (IL1-M52H7) is expressed from human 293 cel ls (HEK293). It contains AA Ala 26 - Thr 340 (Accession NP598751).This protein carries a polyhistidine tag at the C- 1 * 100 µG ACRBIL1-M52H7100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou