MABSol Biotinylated Human Mesothelin (296-580), Fc Tag (MSN-H826x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the biotinylated form of Human Mesothelin (296-580), Fc Tag (MSN-H526x). null
Fournisseur: ACROBIOSYSTEMS
MABSol Biotinylated Human Mesothelin (296-580), Fc Tag (MSN-H826x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the biotinylated form of Human Mesothelin (296-580), Fc Tag (MSN-H526x). null
COMMANDE
Développer tout / Réduire tout
MABSol® Biotinylated Human Mesothelin (296-580), Fc Tag (MSN-H826x) is expressed from human HEK293 cells. It contains AA Glu 296 - Gly 580 (Accession AAH09272). It is the biotinylated form of Human Mesothelin (296-580), Fc Tag (MSN-H526x). null ACRBMSN-H826X200UG Each
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou