Recombinat Protein,Human Niemann-Pick Type C2 NPC2 Protein, His Tag,Human NPC2, His Tag (NP2-H52H1) is expressed from human 293 cel ls (HEK293). It contains AA Glu 20 - Leu 151 (Accession AAH02532).This protein carries a polyhistidine tag at the C-t 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Niemann-Pick Type C2 NPC2 Protein, His Tag,Human NPC2, His Tag (NP2-H52H1) is expressed from human 293 cel ls (HEK293). It contains AA Glu 20 - Leu 151 (Accession AAH02532).This protein carries a polyhistidine tag at the C-t 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Niemann-Pick Type C2 NPC2 Protein, His Tag,Human NPC2, His Tag (NP2-H52H1) is expressed from human 293 cel ls (HEK293). It contains AA Glu 20 - Leu 151 (Accession AAH02532).This protein carries a polyhistidine tag at the C-t 1 * 1 mg ACRBNP2-H52H1-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou