Human Neuropilin-1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession AAH0 7533). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human Neuropilin-1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession AAH0 7533). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human Neuropilin-1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession AAH0 7533). 1 * 1 mg ACRBNR1-H5228-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou