Recombinat Protein,Rabbit PD-1 PDCD1 Protein, His Tag,Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession G1SUF0).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Rabbit PD-1 PDCD1 Protein, His Tag,Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession G1SUF0).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rabbit PD-1 PDCD1 Protein, His Tag,Rabbit PD-1, His Tag (PD1-R52H0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession G1SUF0).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 100 µG ACRBPD1-R52H0100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou