Recombinat Protein,Human PROCR EPCR CD201 Protein, Fc Tag,Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK29 3). It contains AA Ser 18 - Thr 209 (Accession AAH14451).This protein carries a human IgG1 Fc tag at the C-terminus. as 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human PROCR EPCR CD201 Protein, Fc Tag,Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK29 3). It contains AA Ser 18 - Thr 209 (Accession AAH14451).This protein carries a human IgG1 Fc tag at the C-terminus. as 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PROCR EPCR CD201 Protein, Fc Tag,Human PROCR, Fc Tag (PRR-H5254) is expressed from human 293 cells (HEK29 3). It contains AA Ser 18 - Thr 209 (Accession AAH14451).This protein carries a human IgG1 Fc tag at the C-terminus. as 1 * 1 mg ACRBPRR-H5254-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou