Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human PSMA FOLH1 Protein, His Tag,Human PSMA, His Tag (PSA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1 ).This protein carries a polyhistidine tag at the N-terminus. The p a 1 * 1 mg ACRBPSA-H52H3-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou