Recombinat Protein,Mouse ROR1 Protein, His Tag,Mouse ROR1, His Tag (RO1-M5221) is expressed from human 293 cells (HEK293). It contai ns AA Gln 30 - Glu 403 (Accession Q9Z139).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse ROR1 Protein, His Tag,Mouse ROR1, His Tag (RO1-M5221) is expressed from human 293 cells (HEK293). It contai ns AA Gln 30 - Glu 403 (Accession Q9Z139).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse ROR1 Protein, His Tag,Mouse ROR1, His Tag (RO1-M5221) is expressed from human 293 cells (HEK293). It contai ns AA Gln 30 - Glu 403 (Accession Q9Z139).This protein carries a polyhistidine tag at the C-terminus. The protein ha 1 * 1 mg ACRBRO1-M5221-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou