Recombinat Protein,Human S100A14 Protein, His Tag,Human S100A14, His Tag (S14-H5121) is expressed from E.coli cells. It contains AA Met 1 - His 104 (Accession AAH05019).This protein carries a polyhistidine tag at the C-terminus. The protein has a ca 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human S100A14 Protein, His Tag,Human S100A14, His Tag (S14-H5121) is expressed from E.coli cells. It contains AA Met 1 - His 104 (Accession AAH05019).This protein carries a polyhistidine tag at the C-terminus. The protein has a ca 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human S100A14 Protein, His Tag,Human S100A14, His Tag (S14-H5121) is expressed from E.coli cells. It contains AA Met 1 - His 104 (Accession AAH05019).This protein carries a polyhistidine tag at the C-terminus. The protein has a ca 1 * 100 µG ACRBS14-H5121100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou