Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797 .1). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797 .1). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Mouse TIGIT, His Tag (TIT-M52E6) is expressed from human 293 cells (HEK293). It contains AA Gly 26 - Thr 143 (Accession NP001139797 .1). 1 * 50 µG ACRBTIT-M52E6-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou