Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession P98155-2). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession P98155-2). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human VLDL R, His Tag (VLR-H5227) is expressed from human 293 cells (HEK293). It contains AA Gly 28 - Ser 769 (Accession P98155-2). 1 * 1 mg ACRBVLR-H5227-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou