XL-1 red competent cells
Fournisseur: AGILENT
200129 200129
STRA200129EA 734 EUR InStock
STRA200129 AGLS200129
XL-1 red competent cells
Cellules compétentes
XL-1 Red Competent Cells are ideal for introducing random mutations into a cloned gene of interest.
  • Suitable for generating random mutations within genes lacking selectable or screenable phenotypes
  • Does not require extensive genetic or biochemical manipulations
  • Generate random mutations within a gene that has no selectable or screenable phenotype
XL-1 Red Competent Cells can be used to introduce random mutations in a cloned gene of interest through a highly efficient, rapid, and reproducible method. The method involves propagating the cloned gene into XL-1 Red Competent Cells, an Escherichia coli strain deficient in three of the primary DNA repair pathways.
The mutS, mutD, and mutT mutations were introduced into the XL-1 Red strain using basic bacterial genetics. The random mutation rate in this triple mutant strain was measured to be ~5000-fold higher than that of the wild-type.
Attention: For research use only. Not for use in diagnostic procedures.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité