A77453-96
ANTIA77453-96EA
600
EUR
InStock
ANTIA77453-96
Mouse Anti-TSH Receptor/TSH-R Antibody ELISA Kit
Tests
Tests ELISA
Mouse Anti-TSH Receptor/TSH-R Antibody ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Anti-TSH Receptor/TSH-R Antibody in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 1,563 to 100 ng/ml
- Sensitivity: 0,938 ng/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.