Synonyms:
3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine
B23634.14
B23634.14EA
38.6
EUR
InStock
B23634.14
3-(Aminométhyl)pyridine ≥99%
3-(Aminométhyl)pyridine
Purity: 99+%
CAS No.: 3731-52-0
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 100°(212°F)
Refractive Index: 1.5510
Odour: Amine-like
pH: 11-12
Melting Point: -21°
Boiling Point: 226°
Beilstein Reference: 108056
UN No.: 2735
MDL No.: MFCD00006412