Synonyms:
5-Aminotetrazole monohydrate, 5-Amino-1H-tetrazole monohydrate, Tetrazol-5-amine, 1H-Tetrazol-5-ylamine monohydrate, 1H-Tetrazol-5-amine hydrate, 5-Amino-tetrazole-monohydrate
B24018.22
B24018.22EA
74.9
EUR
InStock
B24018.22
5-Aminotétrazole monohydraté 99%
5-Amino-1H-tetrazole monohydrate
Purity: 99%
CAS No.: 15454-54-3
Molecular Formula: CH3N5.H2O
Formula Weight: 103.08 (85.06anhy)
pH: 3.7
Melting Point: ca 203° dec.
Beilstein Reference: 108872
UN No.: 1325
MDL No.: MFCD00149327