Human MyD88 ELISA kit
Fournisseur: Biorbyt
ORB106601-96
BIRBORB106601-96EA 0 EUR InStock
BIRBORB106601-96
Human MyD88 ELISA kit
Tests Tests ELISA
Human MyD88 ELISA Kit for the quantitative detection of human Myeloid differentiation primary response gene 88 (MYD88).
MYD88 is a protein that, in humans, is encoded by the MYD88 gene. By interspecific backcross mapping, the mouse MyD88 gene was localised to chromosome 9; the human homolog was mapped to 3p22-p21. The human ortholog MYD88 seems to function similarly to mice, however, available evidence suggests that MYD88 is dispensable for human resistance to common viral infections and to all but a few pyogenic bacterial infections, demonstrating a major difference between mouse and human immune responses.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité