ORB106616-96
BIRBORB106616-96EA
0
EUR
InStock
BIRBORB106616-96
Mouse CXCL13 ELISA kit
Tests
Tests ELISA
Mouse CXCL13 ELISA Kit for the quantitative determination of mouse Chemokine (C-X-C motif) 13 (CXCL13).
CXCL13, also known as B lymphocyte chemoattractant (BLC), is a protein that in humans is encoded by the CXCL13 gene. CXCL13 is a small cytokine belonging to the CXC chemokine family, and it was located on sequence maps to 4q21, near the genes encoding most CXC chemokines. In T-lymphocytes, CXCL13 expression is thought to reflect a germinal centre origin of the T-cell. Hence, expression of CXCL13 in T-cell lymphomas, such as Angioimmunoblastic T-cell Lymphoma, is thought to reflect a germinal center origin of the neoplastic T-cells.