Human CA9 ELISA kit
Fournisseur: Biorbyt
ORB138063-96
BIRBORB138063-96EA 0 EUR InStock
BIRBORB138063-96
Human CA9 ELISA kit
Tests Tests ELISA
Human CA9 ELISA Kit is used for quantitative detection of human CA9 in cell culture supernates, serum, plasma (heparin, EDTA, citrate) and urine.
Carbonic anhydrase 9 (CA9/CAIX) is an enzyme that in humans is encoded by the CA9 gene. Carbonic anhydrases (CAs) are a large family of zinc metalloenzymes that catalyse the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid.
This gene is mapped to 9p13.3. CA IX is a transmembrane protein and the only tumour-associated carbonic anhydrase isoenzyme known. It is expressed in all clear-cell renal cell carcinoma, but is not detected in normal kidney or most other normal tissues. It may be involved in cell proliferation and transformation.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité