Human EPCR ELISA kit
Fournisseur: Biorbyt
ORB138076-96
BIRBORB138076-96EA 0 EUR InStock
BIRBORB138076-96
Human EPCR ELISA kit
Tests Tests ELISA
Human EPCR ELISA kit for the quantitative detection of human endothelial protein C receptor (EPCR) in cell culture supernates, serum and plasma (heparin, EDTA).
Endothelial protein C receptor (EPCR) also known as CCCA or PROCR is a protein that in humans is encoded by the PROCR gene. PRPCR gene is mapped to 20q11.2 by radiation hybrid analysis and FISH. The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité