Human ICAM-2 ELISA kit
Fournisseur: Biorbyt
ORB138100-96
BIRBORB138100-96EA 0 EUR InStock
BIRBORB138100-96
Human ICAM-2 ELISA kit
Tests Tests ELISA
Human ICAM-2 ELISA kit for the quantitative detection of human Intercellular adhesion molecule 2 (ICAM-2) in cell culture supernates, tissue homogenates, serum and plasma (heparin, EDTA).
ICAM2 also known as CD102 is an integral membrane protein that has 2 immunoglobulin-like domains. This gene is mapped to 17q23.3. The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité