ORB96781-96
BIRBORB96781-96EA
0
EUR
InStock
BIRBORB96781-96
Mouse CTLA4 ELISA kit
Tests
Tests ELISA
Mouse CTLA4 ELISA Kit is a sandwich ELISA kit for the quantitative detection for mouse CTLA4 (Cytotoxic T-Lymphocyte Antigen 4). CTLA4 is a protein receptor that down regulates the immune system.
- No detectable cross-reactivity with other relevant proteins