Human HBEGF ELISA kit
Fournisseur: Biorbyt
ORB96788-96
BIRBORB96788-96EA 0 EUR InStock
BIRBORB96788-96
Human HBEGF ELISA kit
Tests Tests ELISA
Human HBEGF ELISA Kit is a sandwich ELISA kit for the quantitative detection of human heparin-binding EGF-like growth factor (HBEGF).
  • No detectable cross-reactivity with other relevant proteins
HBEGF, also known as HEGFL or DTR, is a member of the EGF family of proteins that in humans is encoded by the HBEGF gene. The HBEGF gene is assigned to chromosome 5, thus confirming the assignment of the gene on the basis of its role in relation to diphtheria toxin susceptibility. HBEGF is an 87 amino acid glycoprotein which displays highly regulated gene expression. It has been shown to play a role in wound healing, cardiac hypertrophy and heart development and function. HBEGF binding and activation of EGF receptors plays a critical role during cardiac valve tissue development and the maintenance of normal heart function in adults.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité