ORB96805-96
BIRBORB96805-96EA
0
EUR
InStock
BIRBORB96805-96
Human SCGB1A1 ELISA kit
Tests
Tests ELISA
Human SCGB1A1 ELISA Kit for the quantitative detection of human secretoglobin family 1A member 1 (SCGB1A1 or uteroglobin).
- There is no detectable cross-reactivity with other relevant proteins
Secretoglobin family 1A member 1 also known as Uteroglobin is a protein that in humans is encoded by the SCGB1A1 gene. SCGB1A1 is the founding member of the secretoglobin family of small, secreted, disulfide-bridged dimeric proteins found only in mammals. This protein is specifically expressed in Clara cells in the lungs. Numerous studies demonstrated that UG is a multifunctional protein with antiinflammatory or immunomodulatory properties. It inhibits soluble phospholipase A2 activity and binds and perhaps sequesters hydrophobic ligands such as progesterone, retinols, polychlorinated biphenyls, phospholipids, and prostaglandins. In addition to its antiinflammatory activities, UG manifests antichemotactic, antiallergic, antitumorigenic, and embryonic growth-stimulatory activities.