Tubing, PTFE, anti-static, explosion-proof, BOLA
Fournisseur: Bohlender
S1827-40 S1827-60 S1827-64 S1827-50 S1827-68 S1827-30 S1827-26 S1827-10 S1827-42 S1827-74
BOHLS1827-40EA 25.3 EUR InStock
BOHLS1827-40 BOHLS1827-60 BOHLS1827-64 BOHLS1827-50 BOHLS1827-68 BOHLS1827-30 BOHLS1827-26 BOHLS1827-10 BOHLS1827-42 BOHLS1827-74
Tubing, PTFE, anti-static, explosion-proof, BOLA
Tuyaux Transfer Tubing
Electric conductive black tubing with antistatic compound made of pure PTFE and finest, highly pure carbon dust (less than 2,5%). Ideal for light-sensitive substances.
  • Extensive chemical resistance due to PTFE parts
  • Temperature resistance from −200 to +250 °C
  • Resistance of less than 1,000 kOhm
They have antistatic applications, used in explosive ambiance (explosion protection), also for transport of solvents or alcohols.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité