Anti-MYT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Fournisseur: Bioss
BS-3505R-A350
BOSSBS-3505R-A350EA 893 EUR InStock
BOSSBS-3505R-A350
Anti-MYT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorps
Myt1 is a zinc finger protein that is known to interact with the co-repressor Sin3B and also HDAC1 and HDAC2. The Myt1 family, including Myt1 and Myt1L, exemplifies a class of neural sequence specific transcription factors that actively recruit HDACs to selected genes during CNS development.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MYT1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité