Anti-CSTA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Fournisseur: Bioss
BS-5114R-A555
BOSSBS-5114R-A555EA 893 EUR InStock
BOSSBS-5114R-A555
Anti-CSTA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorps
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité