Anti-RAB2 Rabbit Polyclonal Antibody (Cy5®)
Fournisseur: Bioss
BS-6174R-CY5
BOSSBS-6174R-CY5EA 900 EUR InStock
BOSSBS-6174R-CY5
Anti-RAB2 Rabbit Polyclonal Antibody (Cy5®)
Anticorps
The protein encoded by this gene belongs to the Rab family, members of which are small molecular weight guanosine triphosphatases (GTPases) that contain highly conserved domains involved in GTP binding and hydrolysis. The Rabs are membrane-bound proteins, involved in vesicular fusion and trafficking. This protein is a resident of pre-Golgi intermediates, and is required for protein transport from the endoplasmic reticulum (ER) to the Golgi complex. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Recommended Dilutions: IF(IHC-P): 1:50-200More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité