Anti-ANGEL2 Rabbit Polyclonal Antibody (Cy7®)
Fournisseur: Bioss
BS-9742R-CY7
BOSSBS-9742R-CY7EA 893 EUR InStock
BOSSBS-9742R-CY7
Anti-ANGEL2 Rabbit Polyclonal Antibody (Cy7®)
Anticorps
ANGEL2 is a 544 amino acid protein that belongs to the CCR4/nocturin family and exists as two alternatively spliced isoforms. The CCR4 family of proteins are 3′–5′-deadenylases that function in the first step of the degradation of poly(A) mRNA. The CCR4 family most likely displays both RNA and ssDNA substrate preferences, thereby implicating a potential role in many regulatory processes. The ANGEL2 gene maps to human chromosome 1 (1q32.3), which is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. Chromosome 1 contains about 3,000More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité