CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
Fournisseur: Brady
176522
BRDY176522EA 0 EUR InStock
BRDY176522
CR950 Handheld Wired Barcode Scanner: 1D, 2D, QR Code
Systèmes de code-barres
Improve data collection operations with this swift and accurate barcode reader. The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device. This kit features a stand.
  • Swift and accurate barcode reader
  • Kit includes a stand
  • Manual or automatic triggering
  • Provides easier scanning with a wide area image sensor
  • Features two programmable indicators: LED and tone
  • 5-year limited warranty
The CR950 thrives in fast-paced environments, offering increased efficiency with its intuitive targeting device. Get high-speed omnidirectional reading of 1D and 2D barcodes and glare reduction technology for reading barcodes on shiny surfaces.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité