Anti-ABR Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9972
BSBTPB9972EA 552 EUR InStock
BSBTPB9972
Anti-ABR Rabbit Polyclonal Antibody
Anticorps
This ABR gene encodes a protein that is similar to the protein encoded by the breakpoint cluster region gene located on chromosome 22. The protein encoded by this gene contains a GTPase-activating protein domain, a domain found in members of the Rho family of GTP-binding proteins. Functional studies in mice determined that this protein plays a role in vestibular morphogenesis. Alternatively spliced transcript variants have been reported for this gene.
1. Heisterkamp, N., Kaartinen, V., van Soest, S., Bokoch, G. M., Groffen, J. Human ABR encodes a protein with GAP-rac activity and homology to the DBL nucleotide exchange factor domain. J. Biol. Chem. 268: 16903-16906, 1993.
2. Heisterkamp, N., Morris, C., Groffen, J. ABR, an active BCR-related gene. Nucleic Acids Res. 17: 8821-8831, 1989.
3. McDonald, J. D., Daneshvar, L., Willert, J. R., Matsumura, K., Waldman, F., Cogen, P. H. Physical mapping of chromosome 17p13.3 in the region of a putative tumor suppressor gene important in medulloblastoma. Genomics 23: 229-232, 1994.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité