Anti-ACO2 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9973
BSBTPB9973EA 552 EUR InStock
BSBTPB9973
Anti-ACO2 Rabbit Polyclonal Antibody
Anticorps
Aconitase 2, mitochondrial is a protein that in humans is encoded by the ACO2 gene. The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification.
1. "Entrez Gene: Aconitase 2, mitochondrial".
2. Geurts van Kessel, A. H. M., Westerveld, A., de Groot, P. G., Meera Khan, P., Hagemeijer, A. Regional localization of the genes coding for human ACO2, ARSA, and NAGA on chromosome 22. Cytogenet. Cell Genet. 28: 169-172, 1980.
3. Mirel, D. B., Marder, K., Graziano, J., Freyer, G., Zhao, Q., Mayeux, R., Wilhelmsen, K. C. Characterization of the human mitochondrial aconitase gene (ACO2). Gene 213: 205-218, 1998.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité