Anti-ADAR Rabbit Polyclonal Antibody
Fournisseur: BosterBio
PB9976
BSBTPB9976EA 552 EUR InStock
BSBTPB9976
Anti-ADAR Rabbit Polyclonal Antibody
Anticorps
Double-stranded RNA-specific adenosine deaminase, also known as ADAR1, is an enzyme that in humans is encoded by the ADAR gene. It is mapped to 1q21.3. This gene encodes the enzyme responsible for RNA editing by site-specific deamination of adenosines. This enzyme destabilizes double-stranded RNA through conversion of adenosine to inosine. Mutations in this gene have been associated with dyschromatosis symmetrica hereditaria. Alternative splicing results in multiple transcript variants.
1. "Entrez Gene: ADAR Adenosine Deaminase Acting on RNA".
2. Kim U, Wang Y, Sanford T, Zeng Y, Nishikura K (November 1994). "Molecular cloning of cDNA for double-stranded RNA adenosine deaminase, a candidate enzyme for nuclear RNA editing". Proceedings of the National Academy of Sciences of the United States of America 91 (24): 11457–61.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité