Anti-MMP12 Rabbit Polyclonal Antibody
Fournisseur: BosterBio
RP1089
BSBTRP1089EA 552 EUR InStock
BSBTRP1089
Anti-MMP12 Rabbit Polyclonal Antibody
Anticorps
Rabbit IgG polyclonal antibody for Macrophage metalloelastase(MMP12) detection. Tested with WB, ELISA in Mouse.
Matrix metalloproteinase-12 (MMP12), also known as MME or ME, is an enzyme that in humans is encoded by the MMP12 gene. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.2. It is thought that the protein encoded by this gene is cleaved at both ends to yield the active enzyme, but this processing has not been fully described. The enzyme degrades soluble and insoluble elastin. It may play a role in aneurysm formation and studies in mice suggest a role in the development of emphysema. This gene may involved in tissue injury and remodeling.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité