Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Fournisseur: ENTEGRIS
CKM12-12F-12F CKM12-12K-12K
ENTECKM12-12F-12FEA 0 EUR InStock
ENTECKM12-12F-12F ENTECKM12-12K-12K
Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Vannes Check Valves
Corrosion-resistant Galtek® check valves reduce contamination potential in your high-purity chemical applications and provide clean delivery of sophisticated chemicals.
  • O-ring free design reduces potential contamination
  • Simple, clean, compact valve provides high flow chemical delivery
  • No metal parts - will not corrode or contribute to contamination
  • Poppet design allows a high flow through rate and maximizes rinsability
An O-ring-free design provides a simple, clean, compact valve with a high flow rate and increased reliability. All wetted parts areMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité