Synonyms:
Pseudochelerythrine, Pseudochelerythrine chloride
ALX350076M005
ENZOALX350076M005EA
251
EUR
InStock
ENZOALX350076M005
Sanguinarine chloride ≥98% (par HPLC)
Sanguinarine chloride
Na⁺/K⁺- and Mg2⁺-ATPase inhibitor
Na+/K+-and Mg2+-ATPase inhibitor. Antimicrobial, anti-inflammatory and antioxidant properties. Induces apoptosis. Modulates the Bcl-2 family. Shows antiplaque activity in humans.
Isolated from the leaves and stems of Macleaya cordata and M. microcarpa.