AMPIGENE® HS Taq DNA Polymerase
Fournisseur: ENZO LIFE SCIENCES
ENZPRT1010500
ENZOENZPRT1010500EA 334 EUR InStock
ENZOENZPRT1010500
AMPIGENE® HS Taq DNA Polymerase
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Increased sensitivity standard PCR for a broader range of samples, with enhanced speed, yield, and specificity.
AMPIGENE® HS Taq DNA Polymerase uses the latest developments in polymerase technology and buffer chemistry to optimize and increase PCR sensitivity. The AMPIGENE® HS Taq DNA Polymerase and 5x AMPIGENE® reaction buffer are separate for flexibility and end-user customization. Hot-start technology is used to increase sensitivity for PCR applications including genotyping, multiplex PCR, screening, library construction, colony PCR and PCR direct from blood and urine.The AMPIGENE® HS Taq DNA Polymerase is resistant to PCR inhibitors and suitable for direct PCR from unprocessed samples: bacterial culture, bacterial colonies, blood, and urine.
Informations de commande: Contains AMPIGENE® HS Taq DNA polymerase (5u/µl) 5x AMPIGENE® reaction buffer (15mM MgCl2, 5mM dNTPs, enhancers and stabilizers).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité