Duo-Ready DNA Ultra 2.0 prefilled plates, KingFisher™
Fournisseur: Thermo Fisher Scientific
A36584
FINNA36584EA 820 EUR InStock
FINNA36584
Duo-Ready DNA Ultra 2.0 prefilled plates, KingFisher™
Systèmes de purification des acides nucléiques
KingFisher Duo-Ready DNA Ultra 2.0 prefilled plates are automation-ready plates prefilled with MagMAX DNA Multi-Sample Ultra 2.0 reagents suitably configured for KingFisher Duo and KingFisher Duo Prime purification systems. KingFisher Duo-Ready DNA Ultra 2.0 Prefilled Plates enable isolation of genomic DNA from 50 to 400 µl of multiple sample types such as whole blood, saliva, buffy coat, buccal swabs, and more. The purified gDNA is ideal for a variety of downstream molecular biology applications.
  • Maximise convenience with MagMAX DNA Multi-Sample Ultra 2.0 isolation chemistry prefilled into KingFisher SBS microtiter plates
  • Increase ease of use—process 12 samples on instrument in 45 minutes without the hassle of reagent setup
  • Designed to reduce user error
  • Helps to reduce hands-on time
  • Formatted specifically for KingFisher Duo instruments
Contents: 8 KingFisher Duo-Ready DNA Ultra 2.0 Prefilled plates, 4,5 ml proteinase K solution, 4,5 ml enhancer solution, 12 ml elution solution, 8 KingFisher Duo 12-tip combs, 8 KingFisher elution strips and caps, 1 elution strip rack.
Attention: Store at room temperature.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité