These labels, provided in A4 sheet format, are compatible with both desktop laser and inkjet printers. Perfect for general-use applications in labs, offices, and warehouses.
- Temperature range: −29 to 120 °C (−20,2 to 248 °F)
- Compatible with both laser and inkjet printers, for convenient printing
- Will permanently adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal
- High print quality with vivid colours
- Premium materials allow for uniform ink absorption and brightness
- Easy to customise
General-use, high printout quality, matte paper labels for laser and inkjet printers. Ideal for office use on files, folders, boxes, and containers. Strong permanent adhesive will adhere to a variety of surfaces, including paper, plastic, glass, cardboard, wood, and metal.