Synonyms:
p-Toluenesulphonic acid 3-Oxetanyl ester, 3-Oxetanyl p-Toluenesulphonate, 3-Oxetanyl Tosylate
H57569.06
H57569.06EA
534
EUR
InStock
H57569.06
Oxetan-3-yl 4-methylbenzenesulphonate 96%
Oxetan-3-yl 4-methylbenzenesulphonate
Purity: 96%
CAS No.: 26272-83-3
Molecular Formula: C10H12O4S
Formula Weight: 228.27
Flash Point: 179°(354°F)
Boiling Point: 372°
MDL No.: MFCD08544401