Packaged under argon in resealable ChemSeal™ bottles.
H58544.AD
H58544.ADEA
218
EUR
InStock
H58544.AD
2-Pyridylzinc bromide solution 0.5 M dans le THF
2-Pyridylzinc bromide
It is used in the preparation of P,N ligands for asymmetric Ir-catalyzed reactions.
Handle under dry protective gas. Air sensitive. Store away from strong bases and oxidizing agents, heat, acids and water.