Synonyms:
5-Bromo-2-cyanopyrimidine, 5-bromo-pyrimidine-2-carbonitile
H61363.06
H61363.06EA
203
EUR
InStock
H61363.06
5-Bromopyrimidine-2-carbonitrile 95%
5-Bromopyrimidine-2-carbonitrile
Purity: 95%
CAS No.: 38275-57-9
Molecular Formula: C5H2BrN3
Formula Weight: 183.99
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White to yellow
Beilstein Reference: 775324
UN No.: 3439
MDL No.: MFCD02940446