Synonyms:
N-Fmoc-L-proline pentafluorophenyl ester
H63457.06
H63457.06EA
126
EUR
InStock
H63457.06
Fmoc-Pro-OPfp 98%
Fmoc-Pro-OPfp
Purity: 98%
CAS No.: 86060-90-4
Molecular Formula: C26H18F5NO4
Formula Weight: 503.42
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 127-129°
Beilstein Reference: 4731096
MDL No.: MFCD00065672