Specifications:- Guard Column: Yes. Hardware: SS. Inner Diameter (ID): 0.3 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Colum n Maximum Temperature: 60 C . LC Column Phase: C18. Length: 50 mm. Part Number Application: Peptide Mapping. Part Num 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Guard Column: Yes. Hardware: SS. Inner Diameter (ID): 0.3 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Colum n Maximum Temperature: 60 C . LC Column Phase: C18. Length: 50 mm. Part Number Application: Peptide Mapping. Part Num 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Guard Column: Yes. Hardware: SS. Inner Diameter (ID): 0.3 mm. LC Column Brand: AdvanceBio Peptide Mapping. LC Colum n Maximum Temperature: 60 C . LC Column Phase: C18. Length: 50 mm. Part Number Application: Peptide Mapping. Part Num 1 * 1 Pce HEWL5065-9946 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou