Specifications:- Inner Diameter (ID): 0.12 mm. LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compar tment Valve. LC System Compatibility: 1200 LC System 1260 Infinity LC System. LC Use With Instrument: G1316C. Techniq 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Inner Diameter (ID): 0.12 mm. LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compar tment Valve. LC System Compatibility: 1200 LC System 1260 Infinity LC System. LC Use With Instrument: G1316C. Techniq 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Inner Diameter (ID): 0.12 mm. LC Application Compatibility: Analytical. LC Module Type Compatibility: Column Compar tment Valve. LC System Compatibility: 1200 LC System 1260 Infinity LC System. LC Use With Instrument: G1316C. Techniq 1 * 1 Pce HEWL5067-6187 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou