Key design features of nebulisers for PerkinElmer ICP-MS systems provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The GemClean Cross-Flow II with GemTips is optimised for ICP-MS (0,23 and 0,33 mm orifices) and specially manufactured and cleaned for ultratrace analysis. OpalMist PFA, with uptake at 0,4 ml/min, is designed for high-precision analyses requiring high chemical resistance to HF. Alternatively, the MicroMist, micro-uptake nebuliser delivers high performance for small sample volumes.