Specifications:- Needle Inside Diameter (ID): 0.26 mm. Needle Length: 57 mm. Needle Material: Stainless steel. Needle Outside Diamet er (OD): 0.464 mm. Needle Point Style: Cone. Needle Type: Removable. Syringe Needle Gauge: 26. 1 * 1 Pce
Fournisseur: Agilent
Specifications:- Needle Inside Diameter (ID): 0.26 mm. Needle Length: 57 mm. Needle Material: Stainless steel. Needle Outside Diamet er (OD): 0.464 mm. Needle Point Style: Cone. Needle Type: Removable. Syringe Needle Gauge: 26. 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
Specifications:- Needle Inside Diameter (ID): 0.26 mm. Needle Length: 57 mm. Needle Material: Stainless steel. Needle Outside Diamet er (OD): 0.464 mm. Needle Point Style: Cone. Needle Type: Removable. Syringe Needle Gauge: 26. 1 * 1 Pce HEWL8010-1375 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou