ZORBAX RRHD Eclipse Plus Phenyl-Hexyl, 2.1 mm, 1.8 um, Plus C8, 3.0 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specific ations:- Carbon Load: 9 %. Endcapped: Double. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. 1 * 3 Pce
Fournisseur: Agilent
ZORBAX RRHD Eclipse Plus Phenyl-Hexyl, 2.1 mm, 1.8 um, Plus C8, 3.0 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specific ations:- Carbon Load: 9 %. Endcapped: Double. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. 1 * 3 Pce
COMMANDE
Développer tout / Réduire tout
ZORBAX RRHD Eclipse Plus Phenyl-Hexyl, 2.1 mm, 1.8 um, Plus C8, 3.0 mm, 1.8 um, 1200 bar pressure limit, UHPLC guard, 3/pk. Specific ations:- Carbon Load: 9 %. Endcapped: Double. Guard Column: Yes. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. 1 * 3 Pce HEWL821725-932 1 * 3 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou